Cole-Parmer Luer to Hose Barb Fitting, Straight Adapter, Natural Kynar®, Cleanoom Packed, Male Slip Luer x 1/8" ID; 10/PK
Representative image only.
Cole-Parmer Luer to Hose Barb Fitting, Straight Adapter, Natural Kynar®, Cleanoom Packed, Male Slip Luer x 1/8" ID; 10/PK
Cole-Parmer | Item: 5011489
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹2,000.00INR / Pkg of 10
₹2,000.00INR / Pkg of 10
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Connect to a female luer body to form a leak-free fluid path
  • Single barb design tightly “grips” the tubing
  • Biopharma compatible and USP 661 and BPOG compliant
  • Male luer x 1/8" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Quickly and easily transition from soft tubing to a luer connection using these adapters. Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. A single barb on one end of the fitting creates a tight seal to keep the tubing in place. Natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. These robust fittings exhibit good mechanical properties, low moisture absorption, and good machinability.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPVDF
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;USP
  • Luer Connection Type (Port 1)Male Slip Luer
  • Luer Connection Type (Port 2)Hose Barb
  • DescriptionLuer to Hose Barb Fitting, Straight Adapter, Natural Kynar®, Cleanoom Packed, Male Slip Luer x 1/8" ID; 10/PK