Cole-Parmer Luer to Hose Barb Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Male Luer x 5/32" ID; 10/Pk
Representative image only.
Cole-Parmer Luer to Hose Barb Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Male Luer x 5/32" ID; 10/Pk
Cole-Parmer | Item: 5011012
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹1,200.00INR / Pkg of 10
₹1,200.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer Luer to Hose Barb Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Male Luer x 5/32" ID; 10/Pk
Cole-Parmer Luer to Hose Barb Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Male Luer x 5/32" ID; 10/Pk
5011012
₹1,200.00INR / Pkg of 10
+ + =
Total Price 4900.00 ADD 3 ITEMS TO CART
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Forms a leak-free luer connection with a quick simple turn
  • Single barb design tightly “grips” the tubing
  • USP Class VI, EU 2002/72/EC, and RoHS compliant
  • Male luer x 5/32" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Quickly and easily transition from soft tubing to a luer connection using these adapters. Luer fitting is a standardized system comprised of a male taper and a female mating part that provides a leak-free connection. A single barb on one end of the fitting creates a tight seal to keep the tubing in place. A transparent alternative to polycarbonate, CrystalVu™ fittings do not contain any bisphenol-A (BPA) that has been found to be harmful to humans. These fittings have excellent chemical resistance, withstand temperatures up to 180°C (356°F), and can be autoclaved multiple times with no crazing.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 2) (in)5/32
  • Regulatory ApprovalsNon-DEHP;RoHS;Non-Phthalate;USP
  • Luer Connection Type (Port 1)Male Luer
  • Luer Connection Type (Port 2)Hose Barb
  • DescriptionLuer to Hose Barb Fitting, Straight Adapter, CrystalVu™, Cleanroom Packed, Male Luer x 5/32" ID; 10/Pk