Cole-Parmer Liquid Nitrogen Withdrawal Device for Dewars 03773-01, -02, and -03
Representative image only.
Cole-Parmer Liquid Nitrogen Withdrawal Device for Dewars 03773-01, -02, and -03
Cole-Parmer | Item: 0377311
Easily transfer liquid nitrogen from Cole-Parmer® 25, 35, and 50 L storage dewars
₹356,000.00INR / Each
₹356,000.00INR / Each
Product Description
Key Features
  • Connects directly to a special neck flange on dewars
  • Flow rates up to 8 L/min at 7.25 psi
  • 3 month warranty
  • Works with IC Biomedical 25LD, 35LD, and 50LD
More About this Item
Connect the liquid nitrogen withdrawal device to storage dewars 03773-01, -02, or -03 to dispense liquid nitrogen. Withdrawal device clamps directly onto the special neck flange of the dewars—no other tools required. Use the withdrawal device to transfer liquid nitrogen at rates up to 8 L/min at 7.25 psi. Dispense liquid nitrogen directly from the device using the provided spout or through a standard cryogenic hose connected to the outlet. Uniquely designed device allows filling of the storage dewar without removing the device or venting system pressure.
3 Month Warranty
Specifications
  • Compatible WithLiquid nitrogen storage dewars 03773-01, -02, and -03
  • DescriptionLiquid Nitrogen Withdrawal Device for 03773-01, -02, and -03
  • Warranty3 months