Cole-Parmer LCGC High-Recovery Snap Top Vials, Glass, 11 mm Short Thread, 1.5 mL, Pre-Slit PTFE/Silicone Septum; 100/pk
Representative image only.
Cole-Parmer LCGC High-Recovery Snap Top Vials, Glass, 11 mm Short Thread, 1.5 mL, Pre-Slit PTFE/Silicone Septum; 100/pk
Cole-Parmer | Item: 9870163
Decrease your risk of failed analysis and sample reruns
₹26,100.00INR / Pkg of 100
₹26,100.00INR / Pkg of 100
Product Description
Key Features
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
  • High-recovery vials
  • Pre-slit PTFE/silicone septum
More About this Item
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Snap-top vials come with snap caps that provide a secure seal to minimize evaporation, even with volatile samples. An audible click ensures a secure seal has been formed.
Glass vials are made of Type I, Class A 33-expansion borosilicate glass. High-recovery vials deliver maximum sample capacity (1.5 mL) with minimal residual volume (<4 µL)—use with bottom draw-port needles.
Please Note: Vials, caps, and septa formerly under the Kinesis brand have now been rebranded Cole-Parmer. All the same great features and specifications, just with a new and improved look.
Specifications
  • Volume (mL)1.2
  • Tube MaterialGlass
  • Tube ColorClear
  • Cap TypeSnap Top
  • Cap Diameter (mm)11
  • Septum TypePre-slit PTFE/silicone
  • SterileNo
  • Special FeaturesHigh-Recovery
  • DescriptionLCGC High-Recovery Snap Top Vials, Glass, 11 mm Short Thread, 1.5 mL, Pre-Slit PTFE/Silicone Septum; 100/pk