Cole-Parmer LCGC High-Recovery Screw Top Vials, Amber Glass, 9 mm Short Thread, 1.5 mL, Pre-Slit PTFE/Silicone Septum; 100/pk
Representative image only.
Cole-Parmer LCGC High-Recovery Screw Top Vials, Amber Glass, 9 mm Short Thread, 1.5 mL, Pre-Slit PTFE/Silicone Septum; 100/pk
Cole-Parmer | Item: 9870166
Decrease your risk of failed analysis and sample reruns
₹30,900.00INR / Pkg of 100
₹30,900.00INR / Pkg of 100
Product Description
Includes
Screw top caps with septa and certificate of analysis.
Key Features
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
  • High-recovery vial constructed of amber glass
  • Pre-slit PTFE/silicone septum
More About this Item
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Short thread vials have a wide opening (6.0 mm) with a unique 9-mm thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
High-recovery vials deliver maximum sample capacity (1.5 mL) with minimal residual volume (<4 µL)—use with bottom draw-port needles. Clear glass vials are made of Type I, Class A 33-expansion borosilicate glass. Amber vials are made of Type I, Class B 51-expansion borosilicate glass and are ideal for light-sensitive samples.
Please Note: Vials, caps, and septa formerly under the Kinesis brand have now been rebranded Cole-Parmer. All the same great features and specifications, just with a new and improved look.
Specifications
  • Volume (mL)1.5
  • Tube MaterialGlass
  • Tube ColorAmber
  • Cap TypeShort Thread Screw
  • Cap Diameter (mm)9
  • Septum TypePre-slit PTFE/silicone
  • SterileNo
  • Special FeaturesHigh-Recovery
  • DescriptionLCGC High-Recovery Screw Top Vials, Amber Glass, 9 mm Short Thread, 1.5 mL, Pre-Slit PTFE/Silicone Septum; 100/pk