Cole-Parmer Laboratory Sample Labeling Kit
Representative image only.
Cole-Parmer Laboratory Sample Labeling Kit
Cole-Parmer | Item: 2500031
Labeling tape and markers combined to save money
₹19,000.00INR / Pkg of 1
₹19,000.00INR / Pkg of 1
Buy It Together
Current Item
Cole-Parmer Laboratory Sample Labeling Kit
Cole-Parmer Laboratory Sample Labeling Kit
2500031
₹19,000.00INR / Pkg of 1
+ =
Total Price 21900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
twenty-four rolls of assorted colored tape (3 blue, 3 green, 3 orange, 3 red, 3 pink, 5 white, and 4 yellow), and six black permanent markers.
Key Features
  • Convenient kit includes twenty-four rolls of tape and six black markers
  • Save money and color code your samples — tape comes in assorted colors
More About this Item
Durable labeling tape is waterproof, and oil and acid resistant. Each roll is 14 yards (12.8 m) long and 1/2" (12.7 mm) wide, and withstands temperatures from –10 to 250ºF (–23 to 121ºC). The permanent markers are also waterproof, and the nonfading ink dries instantly. The extra-fine 0.3 mm writing tip stays sharp and precise.
Specifications
  • Tip StyleExtra fine
  • Pen ColorBlack
  • Max Temperature (° F)250
  • Max Temperature (° C)121
  • DescriptionLaboratory Sample Labeling Kit