Cole-Parmer Lab-Type ATC Probe, 100 Ohms RTD
Representative image only.
Cole-Parmer Lab-Type ATC Probe, 100 Ohms RTD
Cole-Parmer | Item: 0599440
Optimize your system with temperature compensation
₹14,500.00INR / Each
₹14,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer Lab-Type ATC Probe, 100 Ohms RTD
Cole-Parmer Lab-Type ATC Probe, 100 Ohms RTD
0599440
₹14,500.00INR / Each
+ =
Total Price 24700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed with a durable epoxy body
  • 1 year warranty
More About this Item
Provides ATC (automatic temperature compensation) for meters with a miniature audio plug connection.
1 Year Warranty
Specifications
  • DescriptionLab-Type ATC Probe, 100 Ohms RTD