Cole-Parmer "L" Clip for RMC Freezer Racks
Representative image only.
Cole-Parmer "L" Clip for RMC Freezer Racks
Cole-Parmer | Item: 2441028
Customize or upgrade your freezer rack
₹1,300.00INR / Each
₹1,300.00INR / Each
Product Description
Key Features
  • Constructed of durable stainless steel
More About this Item
Suitable for use with your PolarSafe freezer racks.
These clips are removable and designed to be adjusted so that the adjustable upright freezer series racks can hold both standard 2” and 3” boxes. These clips are compatible with the adjustable upright freezer models 04404-77, 04404-78, 04404-79, 04404-80, 04404-81, 04404-82, 04404-83, 04404-84, and 04404-85.
L-Clip is suitable for top and bottom shelves while the T-Clip is for the middle shelf.
Specifications
  • MaterialStainless Steel
  • Compatible WithAdjustable upright freezer
  • Description"L" Clip for RMC Freezer Racks