Cole-Parmer KIT 2 Photometric Accuracy Calibration Kit for Spectroscopy
Representative image only.
Cole-Parmer KIT 2 Photometric Accuracy Calibration Kit for Spectroscopy
Cole-Parmer | Item: 0266002
Easily and accurately perform certified spectrophotometer evaluations in any visible or UV-visible spectrophotometer!
₹870,000.00INR / Each
₹870,000.00INR / Each
Product Description
Includes
one hard plastic carrying case.
Key Features
  • Standards are durable, scratch-resistant and never need to be replaced
  • ISO 17025 accredited for testing and calibration and NIST compatible
  • 1 year warranty
More About this Item
Verify the performance and accuracy of your spectrophotometer right in your own lab. These permanent standards do not have to be recalibrated due to material aging. Unique material construction is scratch resistant and provides stable optical performance.
Photometric Accuracy Calibration Kit 02660-02—use for photometric accuracy and absorbance accuracy between 400 and 700 nm.
1 Year Warranty
Specifications
  • Stray LightNo
  • Photometric AccuracyYes
  • DescriptionPhotometric Accuracy Calibration Kit for Spectroscopy