Cole-Parmer Kettle Clamp, Nickel-Plated Zinc, Fits 500 to 1000 mL
Representative image only.
Cole-Parmer Kettle Clamp, Nickel-Plated Zinc, Fits 500 to 1000 mL
Cole-Parmer | Item: 0804750
Provide support for your specific laboratory equipment
₹84,800.00INR / Each
₹84,800.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Designed for use with reaction kettles to holds both the flask and cover firmly together, includes insulated spring-activated clamping arms.
1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc
  • Clamp TypeEquipment, Reaction Kettle
  • Max Grip Range (InchShort)4.92
  • DescriptionKettle Clamp, Nickel-Plated Zinc, Fits 500 to 1000 Ml