Cole-Parmer J30-1-304 Thermocouple, Wire, Type-J, 30-Gauge, Glass Braid Insulation, 1000-ft, Spool
Representative image only.
Cole-Parmer J30-1-304 Thermocouple, Wire, Type-J, 30-Gauge, Glass Braid Insulation, 1000-ft, Spool
Cole-Parmer | Item: 0854201
Make your own extension cables
₹152,000.00INR / Each
₹152,000.00INR / Each
Product Description
Key Features
  • 6 month warranty
More About this Item
Wire comes in 30 gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with fluorinated ethylene propylene resin (FEP/FEP) or fiberglass materials as well as in 100-ft or 1000-ft lengths for any application.
6 Month Warranty
Specifications
  • Connector TypeJ
  • MaterialFiberglass
  • Wire Gauge (Ga)30
  • Error Limit±4°F (±2.2°C) from 32 to 545°F (0 to 285°C); ±0.75% reading from 545 to 1400°F (285 to 750°C)
  • Inner InsulationFiberglass Braid
  • Outer InsulationFiberglass Braid
  • Stranded / SolidSolid
  • Min Temperature (° F)-100
  • Max Temperature (° F)900
  • Min Temperature (° C)-73
  • Max Temperature (° C)482
  • Length (ft)1000
  • Size (in)0.033 in x 0.054 in
  • DescriptionThermocouple, Wire, Type-J, 30-Gauge, Glass Braid Insulation, 1000-ft, Spool