Cole-Parmer ISE Electrode; Fluoride
Representative image only.
Cole-Parmer ISE Electrode; Fluoride
Cole-Parmer | Item: 2707706
Double-junction electrodes easily connect to controller with stripped wire ends
₹57,500.00INR / Each
₹57,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer ISE Electrode; Fluoride
Cole-Parmer ISE Electrode; Fluoride
2707706
₹57,500.00INR / Each
+
Cole-Parmer Fluoride (F-) ISE Double junction solution kits
2750269
₹22,400.00INR / Each
+
Cole-Parmer Replaceable Membrane ISE Probe; Ammonium
2750500
₹64,600.00INR / Each
+ +
Cole-Parmer Ion Concentration Controller
5610500
₹194,000.00INR / Each
=
Total Price 497500.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Compatible with ISE controller 56105-00
  • 1 year warranty
More About this Item
These industrial sealed double-junction electrodes easily connect to the ion selective controller. Industrial electrodes will withstand the conditions of your in-line application. Electrodes measure 12 mm dia x 80 mm L with 10-ft cord.
1 Year Warranty
Specifications
  • Ion-Selective ElectrodeFluoride
  • ConnectorTinned ends
  • DescriptionISE Electrode; Fluoride