Cole-Parmer Irreversible Thermostrip Disinfection Indicator, 160-180F/71-82C; 16/Pk
Representative image only.
Cole-Parmer Irreversible Thermostrip Disinfection Indicator, 160-180F/71-82C; 16/Pk
Cole-Parmer | Item: 9030812
Provide proof of HACCP compliance that sanitizing temperature has been reached during dishwashing
₹4,600.00INR / Pkg of 16
₹4,600.00INR / Pkg of 16
Product Description
Key Features
  • Waterproof—designed to withstand the rigors of a full wash
More About this Item
Thermostrip labels are self-adhering and waterproof color-changing labels that measure the surface temperature of dishware in dishwashing machines. Label indicator will turn from white to black when surface temperature reaches rated temperature, providing reliable proof that the items being washed have been fully disinfected. Helps hygiene managers demonstrate that safety disinfection standards have been met.
To use, attach label to the thickest clean dry dish, place in dishwasher, and run. Remove and retain label as a permanent record of temperature attained as an excellent evidencebase for HACCP. Accuracy is ±1.8°F (±1°C). Shelf life: one year after date of purchase.
Specifications
  • Number of Temperature Indicators3
  • Max temperature (° F)180
  • Max temperature (° C)82
  • Temperature Points (° F)160, 170, 180
  • Temperature Points (° C)71, 77, 82
  • Initial colorWhite
  • First color changeBlack
  • Height (in)1 3/4
  • Width (in)1
  • DescriptionIrreversible Thermostrip Disinfection Indicator, 160-180F/71-82C; 16/Pk