Cole-Parmer Irreversible 1-Point Square Temperature Label, 108F/42C; 50/Pk
Representative image only.
Cole-Parmer Irreversible 1-Point Square Temperature Label, 108F/42C; 50/Pk
Cole-Parmer | Item: 0806835
Reliable proof that the product was maintained at the correct temperature
₹5,600.00INR / Pkg of 50
₹5,600.00INR / Pkg of 50
Product Description
Key Features
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
  • 2 year warranty
More About this Item
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8°F below 212°F and ±1°F above 212°F.
Two year warranty
Specifications
  • Number of Temperature Indicators1
  • Temperature Points (° F)108
  • Temperature Points (° C)42
  • Initial colorWhite
  • First color changeBlack
  • Height (in)1/2
  • Width (in)1/2
  • DescriptionIrreversible 1-Point Square Temperature Label, 108F/42C; 50/Pk