Cole-Parmer Interchangeable Cable Refillable SJ Glass pH Probe; 210x6mm
Representative image only.
Cole-Parmer Interchangeable Cable Refillable SJ Glass pH Probe; 210x6mm
Cole-Parmer | Item: 0565825
Save space and money by using one cable with multiple electrodes or vice versa
₹30,700.00INR / Each
₹30,700.00INR / Each
Buy It Together
Current Item
Cole-Parmer Interchangeable Cable Refillable SJ Glass pH Probe; 210x6mm
Cole-Parmer Interchangeable Cable Refillable SJ Glass pH Probe; 210x6mm
0565825
₹30,700.00INR / Each
+ =
Total Price 40700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Cable cap rotates independently and will not bunch up
  • 6 month warranty
More About this Item
Match these combination electrodes with one of the connector cables listed at the right. If you use several pH meters that each have different connector types, these electrodes and cables can save you money. The probe-cable interface is moisture proof and noise free, with a sealing O-ring, gold-plated contacts, and PTFE insulation.
6 Month Warranty
Specifications
  • ApplicationsSmall test tubes
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference Electrolyte3.8M KCl with AgCl
  • Max Temperature (° C)100
  • ConnectorDepends upon cable purchased separately: BNC, US standard, or SN6 to 57
  • Diameter (mm)6
  • Length (mm)210
  • DescriptionInterchangeable Cable Refillable SJ Glass pH Probe; 210x6mm
  • Warranty6 month warranty