Cole-Parmer Interchangeable Cable pH Probe, Refillable/SJ/Glass; 154x12mm
Representative image only.
Cole-Parmer Interchangeable Cable pH Probe, Refillable/SJ/Glass; 154x12mm
Cole-Parmer | Item: 0565815
Save space and money by using one cable with multiple electrodes or vice versa
₹27,200.00INR / Each
₹27,200.00INR / Each
Buy It Together
Current Item
Cole-Parmer Interchangeable Cable pH Probe, Refillable/SJ/Glass; 154x12mm
Cole-Parmer Interchangeable Cable pH Probe, Refillable/SJ/Glass; 154x12mm
0565815
₹27,200.00INR / Each
+ =
Total Price 37200.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Cable cap rotates independently and will not bunch up
  • 6 month warranty
More About this Item
Match these combination electrodes with one of the connector cables listed at the right. If you use several pH meters that each have different connector types, these electrodes and cables can save you money. The probe-cable interface is moisture proof and noise free, with a sealing O-ring, gold-plated contacts, and PTFE insulation.
6 Month Warranty
Specifications
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference Electrolyte3.8M KCl with AgCl
  • Max Temperature (° C)100
  • ConnectorDepends upon cable purchased separately: BNC, US standard, or SN6 to 57
  • Diameter (mm)12
  • Length (mm)154
  • DescriptionInterchangeable Cable pH Probe, Refillable/SJ/Glass; 154x12mm
  • Warranty6 month warranty