Cole-Parmer Insertion/Submersion pH Probe; SJ/CPVC/1khm RTD; Spade/BNC
Representative image only.
Cole-Parmer Insertion/Submersion pH Probe; SJ/CPVC/1khm RTD; Spade/BNC
Cole-Parmer | Item: 2730024
Reduce bulb damage with probe guard
₹31,100.00INR / Each
₹31,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer Insertion/Submersion pH Probe; SJ/CPVC/1khm RTD; Spade/BNC
Cole-Parmer Insertion/Submersion pH Probe; SJ/CPVC/1khm RTD; Spade/BNC
2730024
₹31,100.00INR / Each
+
Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Transmitter
0580214
₹46,300.00INR / Each
+ =
Total Price 159700.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Versatile design—threads on both ends for submersion or in-line use
  • Double-junction electrodes available; resist contamination from heavy metals, organics, or AgCI reactive compounds
  • 6 month warranty
More About this Item
In-line or submersible electrodes with probe guard operate from 23 to 176°>F (-5 to 80°>C) and have a PVC housing with a 3/4'' NPT(M) fitting on both ends. Epoxy body withstands pressures to 100 psi. Measure pH from 0 to 14. All electrodes measure 6-3/4''L x 1'' OD, with a 1-3/4'' insertion depth.
Note: 100Ω RTD ATC elements are platinum and conform to DIN curve 43760. ATC connector is miniature audio plug.
Connector for the 1 kΩ RTD and the 3 kΩ Balco ATC elements is a spade lug.
Connector for 100 kΩ thermistor ATC elements is a standard 1/4" phone plug.
Keep a Spare6 Month Warranty
Specifications
  • Electrode TypeSingle junction
  • Electrode Body MaterialPVC
  • Min pH (pH)14
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • ATC ConnectorSpade lugs
  • ATC Element1 kΩ Pt RTD
  • ConnectorBNC
  • DescriptionInsertion/Submersion pH Probe; SJ/CPVC/1khm RTD; Spade/BNC
  • Warranty6 month warranty