Cole-Parmer IN-LINE PH ELECTRODE
Representative image only.
Cole-Parmer IN-LINE PH ELECTRODE
Cole-Parmer | Item: 1594658
No more difficult installations
₹48,000.00INR / Each
₹48,000.00INR / Each
Product Description
Key Features
  • Ideal for high temperature and pressure applications
  • Flat surface prevents build-ups and ensures clean surface for longer life
  • Double-junction design for difficult applications
More About this Item
Use with most meters and controllers - standard BNC connectors and ATC element work with a variety of units|pH range – 0 to 14|Specifications:|Thread size : 3/4” or 1” NPT(M) front and 3/4” NPT rear facing connections|Insertion Depth: 2.4 cm |Housing length: 15 cm|Housing material: Ryton PPS |Temperature range: Up to 100 Degree C|Cable: 10 ft (3 m) with BNC pH connector and tinned-ends ATC connector.
Specifications
  • Electrode TypeIn-Line
  • Electrode Body MaterialRyton PPS
  • Min pH (pH)0
  • Max pH (pH)14
  • Max Temperature (° C)100
  • Thread Size NPT(M) (in)1
  • Thread Size NPT(F) (in)3/4
  • ConnectorStandard BNC
  • Cable Length (m)3
  • DescriptionCole-Parmer IN-LINE PH ELECTRODE