Cole-Parmer Hosecock Clamp, Regular, Stainless Steel
Representative image only.
Cole-Parmer Hosecock Clamp, Regular, Stainless Steel
Cole-Parmer | Item: 0812986
₹23,800.00INR / Each
₹23,800.00INR / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
Hosecocks have a convex clamping surface and rounded edges that prevent accidental cuts and tears of tubing. Adjustment screw with an oversized head provides one-handed flow regulation.
Two year warranty
Specifications
  • Clamp MaterialStainless Steel
  • Clamp TypeTubing
  • Min Grip Range (InchShort)0
  • Max Grip Range (InchShort)0.67
  • Length (in)1.46
  • DescriptionHosecock Clamp, Regular, Stainless Steel