Cole-Parmer Hosecock Clamp, Regular, Nickel-Plated Zinc, 0 to 2/3" Dia Tubing
Representative image only.
Cole-Parmer Hosecock Clamp, Regular, Nickel-Plated Zinc, 0 to 2/3" Dia Tubing
Cole-Parmer | Item: 0802800
₹5,300.00INR / Each
₹5,300.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Hosecocks have a convex clamping surface and rounded edges that prevent accidental cuts and tears of tubing. Adjustment screw with an oversized head provides one-handed flow regulation.
1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc
  • Clamp TypeTubing
  • Min Grip Range (InchShort)0
  • Max Grip Range (InchShort)0.67
  • DescriptionHosecock Clamp, Regular, Nickel-Plated Zinc, 0 to 2/3" Dia Tubing