Cole-Parmer Hose Barb Fitting, Straight Reducer, White Nylon, Cleanroom Packed, 1/4" ID x 1/8" ID; 10/Pk
Representative image only.
Cole-Parmer Hose Barb Fitting, Straight Reducer, White Nylon, Cleanroom Packed, 1/4" ID x 1/8" ID; 10/Pk
Cole-Parmer | Item: 5010897
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
₹3,400.00INR / Pkg of 10
₹3,400.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer Hose Barb Fitting, Straight Reducer, White Nylon, Cleanroom Packed, 1/4" ID x 1/8" ID; 10/Pk
Cole-Parmer Hose Barb Fitting, Straight Reducer, White Nylon, Cleanroom Packed, 1/4" ID x 1/8" ID; 10/Pk
5010897
₹3,400.00INR / Pkg of 10
+ =
Total Price 3610.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • FDA and is BSE/TSE and RoHS compliant
  • 1/4" hose barb x 1/8" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
Easily connect two different size tubing with straight reducers. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These nylon fittings are made from the same resin as natural nylon but with FDA-compliant white coloring. They offer good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Certificate of Compliance
Specifications
  • Fitting MaterialNylon
  • Sterilization MethodE-beam/gamma irradiation, EtO, or autoclave up to 121°C
  • Connection TypeHose Barb Reducer
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Flow PatternStraight
  • ColorWhite
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;RoHS;FDA
  • DescriptionHose Barb Fitting, Straight Reducer, White Nylon, Cleanroom Packed, 1/4" ID x 1/8" ID; 10/Pk