Cole-Parmer Hose Barb Fitting, Straight Reducer, Natural Polycarbonate, 1/4" ID x 1/8" ID; 10/Pk
Representative image only.
Cole-Parmer Hose Barb Fitting, Straight Reducer, Natural Polycarbonate, 1/4" ID x 1/8" ID; 10/Pk
Cole-Parmer | Item: 5014023
Easily connect two different size tubing
₹920.00INR / Pkg of 10
₹920.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer Hose Barb Fitting, Straight Reducer, Natural Polycarbonate, 1/4" ID x 1/8" ID; 10/Pk
Cole-Parmer Hose Barb Fitting, Straight Reducer, Natural Polycarbonate, 1/4" ID x 1/8" ID; 10/Pk
5014023
₹920.00INR / Pkg of 10
+ + + + =
Total Price 11650.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Sharp back-barb securely holds tubing in place
  • Transparent fittiings—easily view fluid flow
  • Meets FDA 21 CFR 177.1580 and NSF 51 requirements, DEHP free, and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • 1/4" hose barb x 1/8" hose barb
  • Manufacturer defect warranty
More About this Item
Create a smooth, in-line transition between different tubing sizes using straight reducers. A specially designed sharp back-barb expands tubing 40 to 50% above its normal ID for a leak-free, clamp-free seal up to 125 psi. Transparent fittings make it easy to verify fluid flow.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
ISO 9001:2008 Certified Supplier
Specifications
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Max Temperature (° F)797
  • Max Temperature (° C)425
  • Regulatory ApprovalsNon-DEHP;NSF-51;FDA;RoHS
  • DescriptionHose Barb Fitting, Straight Reducer, Natural Polycarbonate, 1/4" hose barb x 1/8" hose barb; 10/Pk
  • WarrantyManufacturer defect