Cole-Parmer Hose Barb Fitting, Elbow, Natural Polycarbonate, 1/8" ID; 10/Pk
Representative image only.
Cole-Parmer Hose Barb Fitting, Elbow, Natural Polycarbonate, 1/8" ID; 10/Pk
Cole-Parmer | Item: 5013983
Easily change the flow direction with 90° elbows
₹1,100.00INR / Pkg of 10
₹1,100.00INR / Pkg of 10
Product Description
Key Features
  • Sharp back-barb securely holds tubing in place
  • Transparent fittiings—easily view fluid flow
  • Meets FDA 21 CFR 177.1580 and NSF 51 requirements, DEHP free, and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • 1/8" hose barb
  • Manufacturer defect warranty
More About this Item
The elbow barbed fittings offer precise 90° turns, optimizing layouts in compact systems and preventing kinks. A specially designed sharp back-barb expands tubing 40 to 50% above its normal ID for a leak-free, clamp-free seal up to 125 psi. Transparent fittings make it easy to verify fluid flow.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
ISO 9001:2008 Certified Supplier
Specifications
  • Fitting MaterialPolycarbonate
  • Sterilization MethodEtO or gamma irradiation
  • Connection TypeHose Barb Elbow
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • Barb Size / Tubing Id (Port 2) (in)1/8
  • Flow Pattern90° elbow
  • ColorNatural
  • Max Temperature (° F)797
  • Max Temperature (° C)425
  • Regulatory ApprovalsNSF-51;RoHS;FDA;Non-DEHP
  • DescriptionHose Barb Fitting, Elbow, Natural Polycarbonate, 1/8" Hose Barb; 10/Pk
  • WarrantyManufacturer defect