Cole-Parmer High-Purity Solid Silicone Stoppers, Euro Size 10D; 10/Pk
Representative image only.
Cole-Parmer High-Purity Solid Silicone Stoppers, Euro Size 10D; 10/Pk
Cole-Parmer | Item: 6299404
Stop particle buildup
₹2,900.00INR / Pkg of 10
₹2,900.00INR / Pkg of 10
Product Description
Key Features
  • Constructed of high-purity silicone to resist particle build-up
  • Autoclavable
  • 6 month warranty
More About this Item
Translucent silicone stoppers are especially suited for procedures demanding high purity, such as tissue culture work and food science applications. Food-grade quality stoppers are nontoxic, chemically inert, and resist particle buildup. Withstand repeated sterilization, as well as temperatures from –76 to 392°F (–60 to 200°C). Peak at 428°F (220°C). European size (DIN no.) is noted on stopper. Autoclavable.
6 Month Warranty
Specifications
  • Standard Size NumberN/A
  • European Size Number10D
  • Top Outside Diameter (OD) (mm)9
  • Bottom Outside Diameter (OD) (mm)5
  • Height (mm)20
  • ColorTranslucent
  • MaterialSilicone
  • DescriptionHigh-Purity Solid Silicone Stoppers, Euro Size 10D; 10/Pk