Cole-Parmer High-purity silicone stopper, size 10 1/2 (54D)
Representative image only.
Cole-Parmer High-purity silicone stopper, size 10 1/2 (54D)
Cole-Parmer | Item: 6299434
Stop particle buildup
₹8,900.00INR / Pkg of 2
₹8,900.00INR / Pkg of 2
Product Description
Key Features
  • Constructed of high-purity silicone to resist particle build-up
  • Autoclavable
  • 6 month warranty
More About this Item
Translucent silicone stoppers are especially suited for procedures demanding high purity, such as tissue culture work and food science applications. Food-grade quality stoppers are nontoxic, chemically inert, and resist particle buildup. Withstand repeated sterilization, as well as temperatures from –76 to 392°F (–60 to 200°C). Peak at 428°F (220°C). European size (DIN no.) is noted on stopper. Autoclavable.
6 Month Warranty
Specifications
  • Standard Size Number10.5
  • European Size Number54D
  • Top Outside Diameter (OD) (mm)55
  • Bottom Outside Diameter (OD) (mm)47
  • Height (mm)40
  • ColorTranslucent
  • MaterialSilicone
  • DescriptionHigh-purity silicone stopper, size 10 1/2 (54D)