Cole-Parmer High Precision pH Electrode, Glass/3 ft Cable; BNC
Representative image only.
Cole-Parmer High Precision pH Electrode, Glass/3 ft Cable; BNC
Cole-Parmer | Item: 1594554
Faster response and increased stability at extreme pH values
₹11,300.00INR / Each
₹11,300.00INR / Each
Product Description
Includes
What’s included: 3.3 ft (1m) cable and BNC connector
Key Features
  • Get accurate, reliable and fast readings for all 5 point calibration needs
  • Unique sensor technology offers stable readings to 0.01pH units in 10 seconds
More About this Item
Glass body pH electrodes are ideal for laboratory use as they perform for higher temperatures. This refillable electrode is long lasting and offers more stability in low-ionic strength samples. Electrode measures from 0 to 14 pH and withstands temperature up to 70°C. Compatible with most brands of pH meters with standard BNC connection
Specifications
  • DescriptionHigh Precision pH Electrode, Glass/3 ft Cable; BNC