Cole-Parmer High-Level Chlorine Test Strips, 0 to 10, 000 ppm; 50/Pk

Representative image only.

Cole-Parmer High-Level Chlorine Test Strips, 0 to 10, 000 ppm; 50/Pk

Cole-Parmer | Item: 1810501

Broad-range test strips handle high-level available chlorine testing up to 10,000 ppm

₹13,200.00INR / Pkg of 50

₹13,200.00INR / Pkg of 50
Product Description

Key Features

  • Great for conformance to CDC surface disinfection recommendations

More About this Item

These strips are perfect for testing the available chlorine in bleach solutions. Check available chlorine at 0, 1000, 2500, 5000, 7500, and 10,000 ppm. Each test takes approximately 30 seconds. Packaged in a vial of 50 strips. Each vial is lined with desiccant to maintain freshness.

Specifications
  • TestChlorine, available
  • Measurement Points0, 1000, 2500, 5000, 7500, 10000 ppm
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionHigh-Level Chlorine Test Strips, 0 to 10, 000 ppm; 50/Pk
Reviews
Q&A