Cole-Parmer HF Resistant Submersible pH Electrode, CPVC
Representative image only.
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC
Cole-Parmer | Item: 2710800
Keep original housing and simply replace the electrode cartridge
₹98,200.00INR / Each
₹98,200.00INR / Each
Buy It Together
Current Item
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC
Cole-Parmer HF Resistant Submersible pH Electrode, CPVC
2710800
₹98,200.00INR / Each
+ =
Total Price 175400.00 ADD 2 ITEMS TO CART
Product Description
Includes
Cartridge electrode, housing with 10' detachable BNC cable, and Viton o-ring.
Key Features
  • Fluid flow across the surface provides a cleaning action that extends the life of the electrode and improves performance
  • Flat measuring surface minimizes electrode coating, scratching, and breakage
  • Usable in HF concentrations of up to 1%
  • 1 year warranty
More About this Item
These electrodes are ideal for solutions containing acidic fluorides (hydrofluoric acid).
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialCPVC
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionHF Resistant Submersible pH Electrode, CPVC