Cole-Parmer Heater Block Kit for Hot Plate or Stirring Hot Plate, Small, 25/50/100 mL
Representative image only.
Cole-Parmer Heater Block Kit for Hot Plate or Stirring Hot Plate, Small, 25/50/100 mL
Cole-Parmer | Item: 0480573
Turn your existing 6" x 6" hot plate into a heating mantle
₹149,000.00INR / Each
₹149,000.00INR / Each
Product Description
Key Features
  • Safest way to heat round-bottom flasks
  • Flexible modular system for a wide range of sizes up to 500 mL
  • Low-well design is perfect for multi-neck flasks
  • 1 year warranty
More About this Item
Designed to heat round-bottom flasks, the heating block system with interchangeable inserts can be used with various Cole-Parmer hot plates and hot plate stirrers with 6" x 6" top plate. The heating block system offers two sizes of base plates (small and medium) which can accommodate up to three block inserts each. A third larger base plate features heat-resistant handles and accepts a single block insert for flasks up to 500 mL. Convenient starter kits feature a base plate with a selection of heating block inserts in various sizes. Chemical- and solvent-resistant anodized block inserts provide a safe method for synthesis of single or multiple reactions and offer equal or better heating performance than silicon oil. The low-well provides excellent temperature transfer without compromising the view of the reaction or risk of cracking the glassware. Each base plate features a hole to house a temperature controller probe.
1 Year Warranty
Specifications
  • DescriptionHeater Block Kit for Hot Plate or Stirring Hot Plate, Small, 25/50/100 mL