Cole-Parmer HDPE Dipping Beaker; 1000 mL

Representative image only.

Cole-Parmer HDPE Dipping Beaker; 1000 mL

Cole-Parmer | Item: 6650106

Versatile sampling components to create your own system

₹12,800.00INR / Each

₹12,800.00INR / Each
Product Description

Key Features

  • All parts are made of FDA-compliant HDPE
  • Dipping beakers, stir blades, and sampling bottles use the same thread — use with any handle length

More About this Item

All dipping beakers, stir blades, and sampling bottles use the same thread to be compatible with all three handle length options. Order the handle and individual dipping beaker, stirring paddle, or sample bottle that suits your needs!

Specifications
  • Capacity (mL)1000
  • MaterialFDA-approved HDPE
  • Compatible With66501-01, -02, or -03 handles
  • DescriptionHDPE Dipping Beaker; 1000 mL
Reviews
Q&A