Cole-Parmer Hand Microtome, 10 µm
Representative image only.
Cole-Parmer Hand Microtome, 10 µm
Cole-Parmer | Item: 7890745
Repeatedly slice micro thin sections as thin as 10 microns
₹19,300.00INR / Each
₹19,300.00INR / Each
Product Description
Includes
tempered steel knife.
Key Features
  • Dial is easily turned to select desired specimen thickness
  • Durable, mechanical operation ensures consistent performance without compromising quality
  • Equipped with a clamping mechanism for securing samples of various shapes and sizes
More About this Item
Cole-Parmer® Essentials hand microtome is designed to slice sections precisely and repeatedly from a sample specimen. Easy to use, this microtome is capable of slicing micro thin sections as thin as 10 µm (0.01 mm). This microtome features a clamping mechanism for securing specimens of various shapes and sizes and a piston screwed into a tube whose free end is fixed into a flat plate stage. The knurled base at the bottom of the instrument moves the piston up or down the central tube when rotated. One full rotation of the base raises the sample up 100 microns (0.1 mm). The specimen is fixed on the flat top of the piston and sections are cut by drawing a razor over the upper flat surface of the instrument.
Specifications
  • Sectioning / Trimming Settings10 µm increments
  • DescriptionHand Microtome, 10 µm