Cole-Parmer H2200-SH Extra Shelf; 10.5" x 8"

Representative image only.

Cole-Parmer H2200-SH Extra Shelf; 10.5" x 8"

Cole-Parmer | Item: 0360804

Optimize perfomance of your mini digital incubator

₹11,800.00INR / Each

₹11,800.00INR / Each
Product Description

More About this Item

Choose from a wide range of accessories including a CO2 regulator and additional shelbed for your mini digital incubator.

Specifications
  • DescriptionExtra Shelf for Mini Digital Incubator; 10.5" x 8"
Reviews
Q&A