Cole-Parmer Grounded Submersible pH Electrode with CPVC Housing
Representative image only.
Cole-Parmer Grounded Submersible pH Electrode with CPVC Housing
Cole-Parmer | Item: 2711100
Ground loop interrupt eliminates electrical interference
₹137,000.00INR / Each
₹137,000.00INR / Each
Product Description
Key Features
  • Fluid flow across the surface provides a cleaning action that extends the life of the electrode and improves performance
  • Flat measuring surface minimizes electrode coating, scratching, and breakage
  • 1 year warranty
More About this Item
This submersible electrode is ideal when your system calibrates outside your tank but gives incorrect readings when installed. Combat interference with a solution ground and internal battery powered ground loop interrupt. Built-in amplifier in electrode cartridge permits long cable runs.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialCPVC
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionGrounded Submersible pH Electrode with CPVC Housing