Cole-Parmer Graphite Straight Ferrules for Packed Columns, 1/16"; 10/Pk
Representative image only.
Cole-Parmer Graphite Straight Ferrules for Packed Columns, 1/16"; 10/Pk
Cole-Parmer | Item: 0199230
Reusable one-piece design
₹9,200.00INR / Pkg of 10
₹9,200.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer Graphite Straight Ferrules for Packed Columns, 1/16"; 10/Pk
Cole-Parmer Graphite Straight Ferrules for Packed Columns, 1/16"; 10/Pk
0199230
₹9,200.00INR / Pkg of 10
+ =
Total Price 18900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of durable graphite
More About this Item
Graphite Ferrules are preferred for capillary columns. Maximum temperature: 450degC. Does not stick to glass and is reusable if not over-tightened. One-piece design – no back ferrule.
Specifications
  • DescriptionGraphite straight ferrules, 1/16"; 10/pack