Cole-Parmer Graphite Short Ferrules Compatible with Agilent Inlets, 1/16" to 0.8 mm; 10/Pk
Representative image only.
Cole-Parmer Graphite Short Ferrules Compatible with Agilent Inlets, 1/16" to 0.8 mm; 10/Pk
Cole-Parmer | Item: 0199859
Reusable one-piece design
₹9,700.00INR / Pkg of 1
₹9,700.00INR / Pkg of 1
Buy It Together
Current Item
Cole-Parmer Graphite Short Ferrules Compatible with Agilent Inlets, 1/16" to 0.8 mm; 10/Pk
Cole-Parmer Graphite Short Ferrules Compatible with Agilent Inlets, 1/16" to 0.8 mm; 10/Pk
0199859
₹9,700.00INR / Pkg of 1
+ =
Total Price 18900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Constructed of durable graphite
  • 1 month warranty
More About this Item
Graphite Ferrules are preferred for capillary columns. Maximum temperature: 450°C. Does not stick to glass and is reusable if not over-tightened. One-piece design – no back ferrule.
1 Month Warranty
Specifications
  • DescriptionGraphite Short Ferrules Compatible with Agilent Inlets, 1/16" to 0.8 mm
  • Warranty1 month