Cole-Parmer Frame Rod Platform, Aluminum, 9-1/2" L x 6-1/2" W
Representative image only.
Cole-Parmer Frame Rod Platform, Aluminum, 9-1/2" L x 6-1/2" W
Cole-Parmer | Item: 0805910
Provide support for your specific laboratory equipment
₹26,900.00INR / Each
₹26,900.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Use to support your labware items from lab fames or support stands. The round plate design features a raised rim to prevent your apparatus from falling, requires an additional connector in order to attach clamps to frames or other apparatus. The rectangular plates include an integral connector.
1 Year Warranty
Specifications
  • Clamp MaterialAluminum
  • Clamp TypeEquipment, Plate
  • Width (in)6.5
  • Length (in)9.5
  • DescriptionFrame Rod Platform, Aluminum, 9-1/2" L x 6-1/2" W