Cole-Parmer Foam Grip Tweezers w/ Precision Sharp, Fine Tips
Representative image only.
Cole-Parmer Foam Grip Tweezers w/ Precision Sharp, Fine Tips
Cole-Parmer | Item: 0738749
Electrically static dissipative, ESD-safe material
₹10,600.00INR / Each
₹10,600.00INR / Each
Product Description
Key Features
  • Soft PVC foam-grip handle provides relief from hand fatigue
  • Antimagnetic and anti-acid stainless steel tips
  • Excellent chemical and wear resistance
  • Limited Lifetime Warranty
More About this Item
Choose the tip style that works for your application.
Limited Lifetime Warranty
Specifications
  • Overall Length (in)5 1/8
  • Overall Length (cm)13
  • MaterialStainless Steel w/ ESD Foam Handles
  • SterileNo
  • DescriptionFoam Grip Tweezers w/ Precision Sharp, Fine Tips
  • WarrantyLimited Lifetime