Cole-Parmer Flow cell, Schedule 80 CPVC, 3/4" x 3/4" FNPT
Representative image only.
Cole-Parmer Flow cell, Schedule 80 CPVC, 3/4" x 3/4" FNPT
Cole-Parmer | Item: 9967321
Preconfigured pH kits make it easy to save; modular design reuses housing and replaces electrode as needed
₹12,800.00INR / Each
₹12,800.00INR / Each
Product Description
Key Features
  • Flat-surface electrode design minimizes electrode damage
  • Self-cleaning feature extends life of electrode
  • Choose from submersible and in-line kits
  • 1 year warranty
More About this Item
Purchase a pre-configured kits to meet your application requirements. Economical alternative replaces cartridge and reuses housing and cables.
These self-cleaning, flat surface electrodes have been enhanced with protection against thermal and chemical breakdown. The double-junction pH reference protects from heavy metals, sulfur, iodine, or proteins.
1 Year Warranty
Specifications
  • DescriptionFlow cell, Schedule 80 CPVC, 3/4" x 3/4" FNPT