Cole-Parmer FGABLE48 Bath Lid, Gabled, for 48 Liter Baths
Representative image only.
Cole-Parmer FGABLE48 Bath Lid, Gabled, for 48 Liter Baths
Cole-Parmer | Item: 1212298
Enhance the performance of your Cole-Parmer® baths and circulators
₹57,800.00INR / Each
₹57,800.00INR / Each
Buy It Together
Current Item
Cole-Parmer FGABLE48 Bath Lid, Gabled, for 48 Liter Baths
Cole-Parmer FGABLE48 Bath Lid, Gabled, for 48 Liter Baths
1212298
₹57,800.00INR / Each
+ =
Total Price 360800.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 3 year warranty
  • Previous part number FGABLE48
More About this Item
Lids, trays, temperature probes, and other accessories for your bath.
Manufactured from stainless steel and available to fit 48 liter baths to help prevent evaporation losses. Gabled lids provide extra working headroom within the bath.
3 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithUnheated water bath 16104-45
  • DescriptionBath Lid, Gabled, for 48 Liter Baths
  • Warranty3 Years