Cole-Parmer FGABLE18 Bath Lid, Gabled, for 18 and 26 Liter Baths
Representative image only.
Cole-Parmer FGABLE18 Bath Lid, Gabled, for 18 and 26 Liter Baths
Cole-Parmer | Item: 1212297
Enhance the performance of your Cole-Parmer® baths and circulators
₹49,000.00INR / Each
₹49,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer FGABLE18 Bath Lid, Gabled, for 18 and 26 Liter Baths
Cole-Parmer FGABLE18 Bath Lid, Gabled, for 18 and 26 Liter Baths
1212297
₹49,000.00INR / Each
+ =
Total Price 352000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
  • Previous part number FGABLE18
More About this Item
Lids, trays, temperature probes, and other accessories for your bath.
Manufactured from stainless steel and available to fit 18 and 26 liter baths to help prevent evaporation losses. Gabled lids provide extra working headroom within the bath.
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithUnheated water baths 16112-01 and 16101-89
  • DescriptionBath Lid, Gabled, for 18 and 26 Liter Baths