Cole-Parmer FCC1 High Temperature Cooling Coil
Representative image only.
Cole-Parmer FCC1 High Temperature Cooling Coil
Cole-Parmer | Item: 1212287
Enhance the performance of your Cole-Parmer® baths and circulators
₹32,100.00INR / Each
₹32,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer FCC1 High Temperature Cooling Coil
Cole-Parmer FCC1 High Temperature Cooling Coil
1212287
₹32,100.00INR / Each
+ =
Total Price 335100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Lids, trays, temperature probes, and other accessories for your bath.
Designed for assisting in cooling a hot bath more rapidly by flowing tap water or chilled liquid through it, this simple coiled tube attaches to the base of all Techne thermoregulators with ease. This cooling coil can be used to cool a bath to 5°C above the cooling liquid supply temperature.
1 Year Warranty
Specifications
  • Compatible WithAll Cole-Parmer IC-300 series immersion circulators
  • DescriptionHigh Temperature Cooling Coil