Cole-Parmer FADJ12 Adjustable Bath Tray, for 12 Liter Baths
Representative image only.
Cole-Parmer FADJ12 Adjustable Bath Tray, for 12 Liter Baths
Cole-Parmer | Item: 1212278
Enhance the performance of your Cole-Parmer® baths and circulators
₹20,500.00INR / Each
₹20,500.00INR / Each
Buy It Together
Current Item
Cole-Parmer FADJ12 Adjustable Bath Tray, for 12 Liter Baths
Cole-Parmer FADJ12 Adjustable Bath Tray, for 12 Liter Baths
1212278
₹20,500.00INR / Each
+ =
Total Price 323500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
  • Previous part number FADJ12
More About this Item
Lids, trays, temperature probes, and other accessories for your bath.
Supported by a ball chain and clip the stainless steel trays can be used to alter the depth of the bath.
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithUnheated water bath 16101-88
  • DescriptionAdjustable Bath Tray, for 12 Liter Baths