Cole-Parmer Extension Cable, 10 ft, BNC/BNC

Representative image only.

Cole-Parmer Extension Cable, 10 ft, BNC/BNC

Cole-Parmer | Item: 3580591

BNC connection

₹5,500.00INR / Each

₹5,500.00INR / Each

Buy It Together

Current Item

Cole-Parmer Extension Cable, 10 ft, BNC/BNC
Cole-Parmer Extension Cable, 10 ft, BNC/BNC
3580591

₹5,500.00INR / Each
+ + =
Total Price 27200.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 10 ft or 25 ft lengths

More About this Item

Cables feature BNC connectors. Select from two different lengths.

Specifications
  • DescriptionExtension Cable, 10 ft, BNC/BNC
Reviews
Q&A