Cole-Parmer ESD-Safe ESD Safe Plastic Tweezer Set of 5 w/ Hard Case
Representative image only.
Cole-Parmer ESD-Safe ESD Safe Plastic Tweezer Set of 5 w/ Hard Case
Cole-Parmer | Item: 0738726
Handle sensitive electronic components safely
₹10,500.00INR / Set of 5
₹10,500.00INR / Set of 5
Buy It Together
Current Item
Cole-Parmer ESD-Safe ESD Safe Plastic Tweezer Set of 5 w/ Hard Case
Cole-Parmer ESD-Safe ESD Safe Plastic Tweezer Set of 5 w/ Hard Case
0738726
₹10,500.00INR / Set of 5
+ =
Total Price 34200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Chemical resistant to oils, grease, fuels, and nonpolar solvents
  • Withstand heat to 226°F (130°C)
  • Limited Lifetime Warranty
More About this Item
ESD-safe plastic tweezers, reinforced with 30% carbon fiber, offer excellent wear and abrasion resistance. Featuring hard rigidity, high tensile and flexural strength, these tweezers are ideal for handling sensitive materials such as electronic components, micro-mechanical parts, glass, and ceramic substrates.
Limited Lifetime Warranty
Specifications
  • Overall Length (in)4 1/2
  • Overall Length (cm)11.5
  • MaterialPolyamide 66 reinforced with 30 wt% carbon fiber
  • SterileNo
  • DescriptionFull Plastic Tweezer Set w/ Hard Case
  • WarrantyLimited Lifetime