Cole-Parmer Electrode Support Clamp, Nickel-Plated Zinc
Representative image only.
Cole-Parmer Electrode Support Clamp, Nickel-Plated Zinc
Cole-Parmer | Item: 0802706
Provide support for your specific laboratory equipment
₹12,900.00INR / Each
₹12,900.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
This clamp is designed to supports your electrode with an electrode holder at the desired level and angle. Integrated connector included.
1 Year Warranty
Specifications
  • Clamp MaterialNickel-plated Zinc
  • Clamp TypeEquipment, Electrode Support
  • Length (in)7
  • DescriptionElectrode Support Clamp, Nickel-Plated Zinc