Cole-Parmer Dust-It, Handheld Air Blower
Representative image only.
Cole-Parmer Dust-It, Handheld Air Blower
Cole-Parmer | Item: 1776614
Environmentally friendly - no aerosols and nonconductive tip
₹2,000.00INR / Each
₹2,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Dust-It, Handheld Air Blower
Cole-Parmer Dust-It, Handheld Air Blower
1776614
₹2,000.00INR / Each
+
Traceable 100-Hour Digital Timer
9446127
₹5,600.00INR / Each
=
Total Price 7600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for delicate, difficult to clean, and inaccessible equipment
  • 1 year warranty
More About this Item
Bulb duster blows away dust, lint, and dirt. It provides gentle-pressure pinpoint jets of air exactly where desired. Varying the amount of compression provides for controlled delivery.
One-way inlet valve at the top stops air from being drawn through the tip. Nonconductive plastic sleeve-tip prevents scratches and allows you to easily clean optical lenses, slides, circuit boards, and cuvets. Size is 2 1/4"x 5" and weighs 2 ounces.
1 Year Warranty
Specifications
  • Length (in)5
  • Width (in)2.25
  • DescriptionDust-It, Handheld Air Blower