Cole-Parmer Dry Block/Incubator Liquid-In-Glass Thermometer; 20 to 100C, 76mm Immersion, Organic Liquid Fill

Representative image only.

Cole-Parmer Dry Block/Incubator Liquid-In-Glass Thermometer; 20 to 100C, 76mm Immersion, Organic Liquid Fill

Cole-Parmer | Item: 0800621

Replace mercury thermometers for a specific application with the same accuracy without any of the hazards

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

Cole-Parmer Dry Block/Incubator Liquid-In-Glass Thermometer; 20 to 100C, 76mm Immersion, Organic Liquid Fill
Cole-Parmer Dry Block/Incubator Liquid-In-Glass Thermometer; 20 to 100C, 76mm Immersion, Organic Liquid Fill
0800621

Call For Price
+ =
Total Price 33100.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Non-mercury liquid
  • 5 year warranty

More About this Item

These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PFA coated for added safety which may cause a slight change in accuracy up to 1 division.

Dry block thermometers - designed for use in all standard heating blocks and water baths with most having arrows indicating critical temperatures.

5 Year Warranty
Specifications
  • Length (")12
  • Min Temperature (° C)20
  • Max Temperature (° C)100
  • Accuracy±1.0°C
  • Division (° C)1
  • Thermometer FillRed Spirit
  • Overall Length (InchShort)12
  • Overall Length (mm)300
  • Immersion Depth (mm)76
  • Stem MaterialWhite-Back Glass
  • DescriptionDry Block/Incubator Liquid-In-Glass Thermometer; 20 to 100C, 76mm Immersion, Organic Liquid Fill
  • Warranty5 years
Documents & Certificates
Reviews
Q&A