Cole-Parmer DRNase Free™ Reagent Spray Bottle Surface Decontaminant; 32oz, 4/PK
Representative image only.
Cole-Parmer DRNase Free™ Reagent Spray Bottle Surface Decontaminant; 32oz, 4/PK
Cole-Parmer | Item: 7890144
Instantly eliminate DNase and RNase from labware and surfaces
₹32,300.00INR / Pkg of 4
₹32,300.00INR / Pkg of 4
Product Description
Key Features
  • Works on glass, plastic, stainless steel, and benchtops
  • Replace carcinogenic chemicals, like DEPC
  • Phosphate free and biodegradable
More About this Item
The DRNase Free reagent surface decontaminant is ideal for items and surfaces used in PCR that cannot be autoclaved. Just spray and rinse. This nontoxic formula doesn’t leave a residue, so it won’t interfere with gel polymerization or enzymatic reactions.
Specifications
  • FormatLiquid spray
  • Volume (Liters)3.8
  • ApplicationsGlass, plastic, stainless steel, and benchtops
  • DescriptionReagent Spray Bottle Surface Decontaminant; 32oz, 4/PK