Cole-Parmer Dip, Stir, and Sample Threaded Handle; 3 ft
Representative image only.
Cole-Parmer Dip, Stir, and Sample Threaded Handle; 3 ft
Cole-Parmer | Item: 6650102
Versatile sampling components to create your own system
₹14,400.00INR / Each
₹14,400.00INR / Each
Product Description
Key Features
  • All parts are made of FDA-compliant HDPE
  • Dipping beakers, stir blades, and sampling bottles use the same thread — use with any handle length
More About this Item
All dipping beakers, stir blades, and sampling bottles use the same thread to be compatible with all three handle length options. Order the handle and individual dipping beaker, stirring paddle, or sample bottle that suits your needs!
Specifications
  • MaterialFDA-approved HDPE
  • Compatible With66501 Series beakers, stir blades, and bottles
  • Length (InchShort)36
  • DescriptionDip, Stir, and Sample Threaded Handle; 3 ft