Cole-Parmer DIGITAL IR VORTEX MIXER
Representative image only.
Cole-Parmer DIGITAL IR VORTEX MIXER
Cole-Parmer | Item: 8657906
Digital control of speed and time for high repeatability
₹40,400.00INR / Each
₹40,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer DIGITAL IR VORTEX MIXER
Cole-Parmer DIGITAL IR VORTEX MIXER
8657906
₹40,400.00INR / Each
+ =
Total Price 103200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Standard pop-off cup for single test tube and a 6-ft (1.8-m) power cord with plug
Key Features
  • Unique infrared sensor drastically reduces repetitive strain on your wrists, or operate in continuous “touch” mode
  • Multi-task versatile solution with adjustable stirring speed - up to 3000 rpm for applications from gentle mixing to vigorous vortexing
  • Quiet operation with soft-start feature and controlled ramping
  • Clear Communication: Bright display and timer speed and time constantly shown, possibility to set the operating time through the timer
  • Ergonomic and highly innovative design with four anti-sliding feet providing excellent stability on the bench, usable on many surfaces
  • 3 year warranty
More About this Item
Cole-Parmer® Digital IR Vortex Mixer operates in continuous or infrared system (IR) mode, with adjustable stirring speed up to
3000 rpm. Its unique infrared sensor detects the presence of the test tube and the instrument automatically starts vibrating.
It drastically reduces repetitive strain on user’s wrist, or operate in continuous “touch” mode. The system is equipped with
a large, clear display which shows elapsed time and speed. The timer on the system can be set up to 999:59 minutes; unit
will shut off when session is completed. It’s polymer/zinc alloy construction is easy to clean and offers excellent chemical
resistance.
Ideal for many applications including mixing of many kind of tubes/containers thanks to the broad range of accessories
3 Year WarrantyConformite Europeenne
Specifications
  • Width (InchShort)5 29/32
  • Height (InchShort)5 1/10
  • Power (VAC)100 to 240
  • Warranty3 Years